siRNA Details
Gene Symbol: HIST1H2AC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003512.3 | NP_003503.1 | 1 | 485 | ||
GenBank mRNA | BC017379.1 | - | 1 | 449 | ||
BC050602.1 | - | 1 | 441 | |||
BC085010.1 | - | 1 | 446 | |||
U90551.1 | - | 1 | 494 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
