siRNA Details
Gene Symbol: HIST1H2AC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC017379.1 | - | 2 | 1432 | ||
BC050602.1 | - | 2 | 1424 | |||
BC085010.1 | - | 2 | 1429 | |||
U90551.1 | - | 2 | 1474 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
