siRNA Details
Gene Symbol: CNOT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008225.2 | NP_001008226.1 | 5 | 812 | ||
NM_001190847.1 | NP_001177776.1 | 5 | 812 | |||
NM_001190848.1 | NP_001177777.1 | 5 | 812 | |||
NM_001190849.1 | NP_001177778.1 | 5 | 812 | |||
NM_001190850.1 | NP_001177779.1 | 5 | 812 | |||
NM_013316.3 | NP_037448.2 | 5 | 812 | |||
XM_017012235.1 | XP_016867724.1 | 5 | 673 | |||
GenBank mRNA | AF091094.1 | - | 2 | 76 | ||
AF180475.1 | - | 4 | 481 | |||
AK074493.1 | - | 5 | 778 | |||
AK074671.1 | - | 2 | 39 | |||
AK300365.1 | - | 5 | 778 | |||
AK314540.1 | - | 5 | 777 | |||
AL389980.1 | - | 5 | 766 | |||
BC035590.1 | - | 5 | 772 | 5576434 | ||
U71267.1 | - | 5 | 762 | |||
U71268.1 | - | 5 | 762 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
