siRNA Details
Gene Symbol: CNOT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008225.2 | NP_001008226.1 | 3 | 662 | ||
NM_001190847.1 | NP_001177776.1 | 3 | 662 | |||
NM_001190848.1 | NP_001177777.1 | 3 | 662 | |||
NM_001190849.1 | NP_001177778.1 | 3 | 662 | |||
NM_001190850.1 | NP_001177779.1 | 3 | 662 | |||
NM_013316.3 | NP_037448.2 | 3 | 662 | |||
XM_017012235.1 | XP_016867724.1 | 3 | 523 | |||
GenBank mRNA | AF180475.1 | - | 2 | 331 | ||
AK074493.1 | - | 3 | 628 | |||
AK300365.1 | - | 3 | 628 | |||
AK314540.1 | - | 3 | 627 | |||
AL389980.1 | - | 3 | 616 | |||
BC035590.1 | - | 3 | 622 | 5576434 | ||
U71267.1 | - | 3 | 612 | |||
U71268.1 | - | 3 | 612 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
