siRNA Details
Gene Symbol: ZNF320 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC064408.1 | - | 1434 |
Gene Symbol: RPS27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030.4 | NP_001021.1 | 1 | 294 | ||
GenBank mRNA | AV763942.1 | - | 1 | 295 | ||
BC002658.2 | - | 1 | 277 | 3607957 | ||
BC070219.1 | - | 295 | 6744779 | |||
CN304370.1 | - | 1 | 290 | |||
L19739.1 | - | 4 | 280 | |||
U57847.1 | - | 4 | 273 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
