siRNA Details
Gene Symbol: ZWINT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005413.1 | NP_001005413.1 | 7 | 601 | ||
NM_007057.3 | NP_008988.2 | 7 | 742 | |||
NM_032997.2 | NP_127490.1 | 7 | 742 | |||
XM_017015605.1 | XP_016871094.1 | 7 | 734 | |||
GenBank mRNA | AF067656.1 | - | 7 | 729 | ||
AK093808.1 | - | 5 | 1043 | |||
AK225509.1 | - | 7 | 728 | |||
AK293416.1 | - | 7 | 731 | |||
AK298148.1 | - | 7 | 597 | |||
BC000411.1 | - | 4 | 354 | |||
BC020979.1 | - | 7 | 729 | 3848873 | ||
BC110399.1 | - | 7 | 741 | 5554405 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
