siRNA Details
Gene Symbol: SLC25A42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321544.1 | NP_001308473.1 | 7 | 679 | ||
NM_178526.4 | NP_848621.2 | 7 | 715 | |||
XM_005259861.3 | XP_005259918.1 | 8 | 809 | |||
XM_011527937.2 | XP_011526239.1 | 7 | 691 | |||
XM_017026670.1 | XP_016882159.1 | 6 | 952 | |||
GenBank mRNA | BC045598.1 | - | 7 | 712 | 4819096 | |
FN356975.1 | - | 7 | 690 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
