siRNA Details
Gene Symbol: SLC25A42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321544.1 | NP_001308473.1 | 8 | 1702 | ||
NM_178526.4 | NP_848621.2 | 8 | 1738 | |||
XM_005259861.3 | XP_005259918.1 | 9 | 1832 | |||
XM_011527937.2 | XP_011526239.1 | 8 | 1714 | |||
XM_017026670.1 | XP_016882159.1 | 7 | 1975 | |||
GenBank mRNA | AF521885.1 | - | 1 | 927 | ||
BC041973.1 | - | 1 | 753 | |||
BC045598.1 | - | 8 | 1735 | 4819096 | ||
FN356975.1 | - | 8 | 1713 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
