siRNA Details
Gene Symbol: HIST1H3G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003534.2 | NP_003525.1 | 1 | 252 | ||
GenBank mRNA | BC012185.1 | - | 1 | 683 | ||
BC079835.1 | - | 1 | 257 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
