siRNA Details
Gene Symbol: TRIM72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008274.3 | NP_001008275.2 | 4, 5 | 994 | ||
XM_017023254.1 | XP_016878743.1 | 3, 4 | 530 | |||
GenBank mRNA | AK090695.1 | - | 3, 4 | 439 | ||
AK131485.1 | - | 4, 5 | 939 | |||
BC033211.1 | - | 4, 5 | 924 | 4906353 | ||
DA058421.1 | - | 3, 4 | 439 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
