siRNA Details
Gene Symbol: TRIM72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008274.3 | NP_001008275.2 | 7 | 1858 | ||
XM_017023254.1 | XP_016878743.1 | 6 | 1394 | |||
GenBank mRNA | AK090695.1 | - | 6, 7 | 1303 | ||
AK131485.1 | - | 7 | 1803 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
