siRNA Details
Gene Symbol: MYH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK310004.1 | - | 1756 |
Gene Symbol: ACTA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100.3 | NP_001091.1 | 7 | 1345 | ||
GenBank mRNA | AK096902.1 | - | 8 | 1238 | ||
AK300697.1 | - | 7 | 1241 | |||
BC012597.1 | - | 7 | 1347 | 4291656 | ||
BX648545.1 | - | 6 | 1074 | |||
J00068.1 | - | 7 | 1343 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
