siRNA Details
Gene Symbol: ACTA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100.3 | NP_001091.1 | 7 | 1479 | ||
GenBank mRNA | AK300697.1 | - | 7 | 1375 | ||
BC012597.1 | - | 7 | 1481 | 4291656 | ||
BX648545.1 | - | 6 | 1208 |
Gene Symbol: MYH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK310004.1 | - | 1890 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
