siRNA Details
Gene Symbol: LRSAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005373.3 | NP_001005373.1 | 13 | 1178 | ||
NM_001005374.3 | NP_001005374.1 | 12 | 883 | |||
NM_001190723.2 | NP_001177652.1 | 13 | 1178 | |||
NM_138361.5 | NP_612370.3 | 12 | 1437 | |||
XM_006717316.3 | XP_006717379.1 | 13 | 1178 | |||
XM_017015283.1 | XP_016870772.1 | 13 | 2177 | |||
XM_017015284.1 | XP_016870773.1 | 13 | 1173 | |||
GenBank mRNA | AK056203.1 | - | 13 | 1160 | ||
AK056305.1 | - | 1 | 23 | |||
AK091589.1 | - | 12 | 1185 | |||
AK226106.1 | - | 13 | 1164 | |||
AY358830.1 | - | 13 | 1143 | |||
BC009239.2 | - | 12 | 883 | 2988664 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
