siRNA Details
Gene Symbol: HMGB4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145205.5 | NP_660206.2 | 2 | 1765 | ||
GenBank mRNA | AK097707.1 | - | 2 | 1766 | ||
AK130859.1 | - | 1 | 184 | |||
AK311985.1 | - | 1 | 151 | |||
BC021180.2 | - | 1 | 135 | 5266418 | ||
BI827031.1 | - | 1 | 124 | |||
DQ207367.1 | - | 1 | 135 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
