siRNA Details
Gene Symbol: CUL4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008895.2 | NP_001008895.1 | 9 | 1008 | ||
NM_001278513.1 | NP_001265442.1 | 9 | 1152 | |||
NM_001278514.1 | NP_001265443.1 | 9 | 820 | |||
NM_003589.2 | NP_003580.1 | 9 | 834 | |||
XM_011537523.2 | XP_011535825.1 | 8 | 1368 | |||
GenBank mRNA | AB012193.1 | - | 3 | 182 | ||
AB178950.1 | - | 9 | 885 | |||
AF077188.1 | - | 9 | 745 | |||
AK296700.1 | - | 1 | 129 | |||
AK314835.1 | - | 9 | 1107 | |||
AL833355.1 | - | 8 | 772 | |||
AY365124.1 | - | 9 | 885 | |||
BC008308.1 | - | 9 | 896 | 3537176 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
