siRNA Details
Gene Symbol: AATK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080395.2 | NP_001073864.2 | 14 | 4614 | ||
NM_004920.2 | NP_004911.2 | 13 | 4392 | |||
XM_006722193.2 | XP_006722256.1 | 14 | 4607 | |||
XM_006722194.2 | XP_006722257.1 | 13 | 4459 | |||
XM_006722195.2 | XP_006722258.1 | 12 | 4401 | |||
XM_006722196.2 | XP_006722259.1 | 14 | 4576 | |||
XM_011525509.2 | XP_011523811.1 | 13 | 6043 | |||
XM_011525510.2 | XP_011523812.1 | 13 | 4419 | |||
XM_011525511.2 | XP_011523813.1 | 9 | 4272 | |||
XM_017025421.1 | XP_016880910.1 | 13 | 4406 | |||
XM_017025422.1 | XP_016880911.1 | 13 | 4565 | |||
GenBank mRNA | AB014541.2 | - | 13 | 4392 | ||
AK094931.1 | - | 6 | 1413 | |||
AK131395.1 | - | 14 | 4033 | |||
AK131529.1 | - | 16 | 4243 | |||
BC047378.1 | - | 4 | 2179 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
