siRNA Details
Gene Symbol: AATK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080395.2 | NP_001073864.2 | 8, 9 | 929 | ||
NM_004920.2 | NP_004911.2 | 7, 8 | 707 | |||
XM_006722193.2 | XP_006722256.1 | 8, 9 | 922 | |||
XM_006722194.2 | XP_006722257.1 | 8, 9 | 924 | |||
XM_006722195.2 | XP_006722258.1 | 6, 7 | 716 | |||
XM_006722196.2 | XP_006722259.1 | 8, 9 | 891 | |||
XM_011525509.2 | XP_011523811.1 | 7, 8 | 2358 | |||
XM_011525510.2 | XP_011523812.1 | 7, 8 | 734 | |||
XM_011525511.2 | XP_011523813.1 | 3, 4 | 587 | |||
XM_017025421.1 | XP_016880910.1 | 7, 8 | 721 | |||
XM_017025422.1 | XP_016880911.1 | 7, 8 | 880 | |||
GenBank mRNA | AB014541.2 | - | 7, 8 | 707 | ||
AK131395.1 | - | 6, 7 | 720 | |||
AK131529.1 | - | 8, 9 | 929 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
