siRNA Details
Gene Symbol: AATK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080395.2 | NP_001073864.2 | 11 | 2905 | ||
NM_004920.2 | NP_004911.2 | 10 | 2683 | |||
XM_006722193.2 | XP_006722256.1 | 11 | 2898 | |||
XM_006722194.2 | XP_006722257.1 | 10 | 2750 | |||
XM_006722195.2 | XP_006722258.1 | 9 | 2692 | |||
XM_006722196.2 | XP_006722259.1 | 11 | 2867 | |||
XM_011525509.2 | XP_011523811.1 | 10 | 4334 | |||
XM_011525510.2 | XP_011523812.1 | 10 | 2710 | |||
XM_011525511.2 | XP_011523813.1 | 6 | 2563 | |||
XM_017025421.1 | XP_016880910.1 | 10 | 2697 | |||
XM_017025422.1 | XP_016880911.1 | 10 | 2856 | |||
GenBank mRNA | AB014541.2 | - | 10 | 2683 | ||
BC047378.1 | - | 1 | 470 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
