siRNA Details
Gene Symbol: HIST2H2AA3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003516.2 | NP_003507.1 | 1 | 407 | ||
GenBank mRNA | AK312163.1 | - | 1 | 406 | ||
BC001629.1 | - | 1 | 392 | |||
BC019308.1 | - | 1 | 392 | |||
BC096705.1 | - | 1 | 354 | 40021705 | ||
BC096739.1 | - | 1 | 354 | 40021707 | ||
BC098171.1 | - | 1 | 354 | 40021706 | ||
CR536525.1 | - | 1 | 354 | |||
CR541872.1 | - | 1 | 354 | |||
L19779.1 | - | 1 | 407 |
Gene Symbol: HIST2H2AA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040874.1 | NP_001035807.1 | 1 | 407 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
