siRNA Details
Gene Symbol: EML1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008707.1 | NP_001008707.1 | 12 | 1279 | ||
NM_004434.2 | NP_004425.2 | 11 | 1222 | |||
XM_005267397.1 | XP_005267454.1 | 12 | 1349 | |||
XM_005267398.3 | XP_005267455.1 | 12 | 1282 | |||
XM_005267399.4 | XP_005267456.2 | 12 | 1321 | |||
XM_005267400.4 | XP_005267457.1 | 11 | 1248 | |||
XM_011536540.2 | XP_011534842.1 | 11 | 1295 | |||
XM_011536541.1 | XP_011534843.1 | 12 | 1362 | |||
XM_011536542.2 | XP_011534844.1 | 12 | 1321 | |||
XM_017021074.1 | XP_016876563.1 | 11 | 1292 | |||
GenBank mRNA | AK023861.1 | - | 2 | 911 | ||
AK126980.1 | - | 13 | 1235 | |||
AK299812.1 | - | 12 | 1558 | |||
AK307925.1 | - | 11 | 1214 | |||
BC032541.1 | - | 11 | 1198 | 5533599 | ||
BC033043.1 | - | 11 | 1222 | 4821202 | ||
BX247979.1 | - | 12 | 1291 | |||
U97018.1 | - | 11 | 1221 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
