siRNA Details
Gene Symbol: SGK223 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080826.2 | NP_001074295.2 | 5 | 4531 | ||
XM_005272369.4 | XP_005272426.2 | 6 | 4654 | |||
XM_005272370.4 | XP_005272427.2 | 6 | 5119 | |||
GenBank mRNA | AF075060.1 | - | 1 | 465 | ||
AK122582.1 | - | 5 | 4377 | |||
AL833872.1 | - | 2 | 1988 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
