siRNA Details
Gene Symbol: MEIS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009813.2 | NP_001009813.1 | 12 | 1526 | ||
NM_001301059.1 | NP_001287988.1 | 12 | 1577 | |||
NM_020160.2 | NP_064545.1 | 12 | 1715 | |||
XM_011527136.2 | XP_011525438.1 | 12 | 1724 | |||
XM_011527138.2 | XP_011525440.1 | 10 | 1574 | |||
XM_011527139.2 | XP_011525441.1 | 12 | 1245 | |||
XM_011527140.1 | XP_011525442.1 | 12 | 1744 | |||
XM_017027011.1 | XP_016882500.1 | 12 | 1775 | |||
GenBank mRNA | AB209455.1 | - | 7 | 755 | ||
AK098710.1 | - | 12 | 1439 | |||
AK289950.1 | - | 12 | 1309 | |||
BC025404.1 | - | 12 | 1478 | 4825964 | ||
BC045741.1 | - | 5 | 667 | |||
BC069251.1 | - | 12 | 1148 | 6211294 | ||
BI522513.1 | - | 5 | 370 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
