siRNA Details
Gene Symbol: MEIS3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301059.1 | NP_001287988.1 | 6 | 920 | ||
NM_020160.2 | NP_064545.1 | 6 | 920 | |||
XM_011527139.2 | XP_011525441.1 | 6 | 588 | |||
XM_011527140.1 | XP_011525442.1 | 6 | 1087 | |||
XM_017027011.1 | XP_016882500.1 | 6 | 1118 | |||
GenBank mRNA | AB209455.1 | - | 1 | 98 | ||
AK098710.1 | - | 6 | 782 | |||
AK289950.1 | - | 6 | 514 | |||
AL537583.3 | - | 6 | 661 | |||
BC025404.1 | - | 6 | 683 | 4825964 | ||
BM783223.1 | - | 4 | 296 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
