siRNA Details
Gene Symbol: ACTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199954.1 | NP_001186883.1 | 6 | 1993 | ||
NM_001614.3 | NP_001605.1 | 6 | 1874 | |||
NR_037688.1 | 7 | 1725 | ||||
GenBank mRNA | BC000292.2 | - | 6 | 1800 | 2819345 | |
BC001920.1 | - | 7 | 1613 | 2820489 | ||
BC004223.2 | - | 6 | 1562 | |||
BC007442.2 | - | 6 | 1794 | 2962953 | ||
BC009544.1 | - | 3 | 1088 | |||
BC009848.2 | - | 6 | 1791 | 3939552 | ||
BC010417.2 | - | 4 | 957 | |||
BC010999.2 | - | 6 | 1793 | 3940861 | ||
BC012050.1 | - | 6 | 1794 | 4554944 | ||
BC015005.1 | - | 7 | 1639 | 4855463 | ||
BC015695.2 | - | 6 | 1796 | 4842665 | ||
BC015779.1 | - | 7 | 1620 | 4856294 | ||
BC017450.1 | - | 5 | 1701 | |||
BC018774.1 | - | 7 | 1644 | 4869628 | ||
BC018861.2 | - | 3 | 1234 | |||
BC023548.2 | - | 4 | 1401 | |||
BC053572.1 | - | 6 | 1787 | 3865767 | ||
BC063495.1 | - | 5 | 1726 | |||
X04098.1 | - | 6 | 1808 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
