siRNA Details
Gene Symbol: ACTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199954.1 | NP_001186883.1 | 6 | 2070 | ||
NM_001614.3 | NP_001605.1 | 6 | 1951 | |||
NR_037688.1 | 7 | 1802 | ||||
GenBank mRNA | BC000292.2 | - | 6 | 1877 | 2819345 | |
BC001920.1 | - | 7 | 1690 | 2820489 | ||
BC004223.2 | - | 6 | 1639 | |||
BC007442.2 | - | 6 | 1871 | 2962953 | ||
BC009544.1 | - | 3 | 1165 | |||
BC009848.2 | - | 6 | 1868 | 3939552 | ||
BC010417.2 | - | 4 | 1034 | |||
BC010999.2 | - | 6 | 1870 | 3940861 | ||
BC012050.1 | - | 6 | 1871 | 4554944 | ||
BC015005.1 | - | 7 | 1716 | 4855463 | ||
BC015695.2 | - | 6 | 1873 | 4842665 | ||
BC015779.1 | - | 7 | 1697 | 4856294 | ||
BC017450.1 | - | 5 | 1778 | |||
BC018774.1 | - | 7 | 1721 | 4869628 | ||
BC018861.2 | - | 3 | 1311 | |||
BC023548.2 | - | 4 | 1478 | |||
BC053572.1 | - | 6 | 1864 | 3865767 | ||
BC063495.1 | - | 5 | 1803 | |||
X04098.1 | - | 6 | 1885 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
