siRNA Details
Gene Symbol: ACTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001199954.1 | NP_001186883.1 | 6 | 1668 | ||
NM_001614.3 | NP_001605.1 | 6 | 1549 | |||
GenBank mRNA | AK291937.1 | - | 6 | 1482 | ||
BC000292.2 | - | 6 | 1475 | 2819345 | ||
BC007442.2 | - | 6 | 1469 | 2962953 | ||
BC009544.1 | - | 3 | 763 | |||
BC009848.2 | - | 6 | 1466 | 3939552 | ||
BC010999.2 | - | 6 | 1468 | 3940861 | ||
BC012050.1 | - | 6 | 1469 | 4554944 | ||
BC015695.2 | - | 6 | 1471 | 4842665 | ||
BC017450.1 | - | 5 | 1376 | |||
BC018861.2 | - | 3 | 909 | |||
BC023548.2 | - | 4 | 1076 | |||
BC053572.1 | - | 6 | 1462 | 3865767 | ||
BC063495.1 | - | 5 | 1401 | |||
EU668331.1 | - | 3 | 981 | |||
M16247.1 | - | 3 | 981 | |||
X04098.1 | - | 6 | 1483 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
