siRNA Details
Gene Symbol: PCLO | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014510.2 | NP_055325.2 | 3 | 2584 | ||
NM_033026.5 | NP_149015.2 | 3 | 2584 | |||
XM_017012006.1 | XP_016867495.1 | 3 | 3859 | |||
XM_017012007.1 | XP_016867496.1 | 3 | 3843 | |||
GenBank mRNA | AK297315.1 | - | 3 | 2587 | ||
Y19188.1 | - | 5 | 2025 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
