siRNA Details
Gene Symbol: PRIM2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000947.4 | NP_000938.2 | 9 | 885 | ||
XM_011514731.2 | XP_011513033.1 | 3 | 2652 | |||
XM_017011004.1 | XP_016866493.1 | 10 | 1077 | |||
XM_017011005.1 | XP_016866494.1 | 10 | 1077 | |||
XM_017011006.1 | XP_016866495.1 | 10 | 1077 | |||
XM_017011007.1 | XP_016866496.1 | 7 | 732 | |||
GenBank mRNA | AK291915.1 | - | 9 | 872 | ||
BC064931.1 | - | 9 | 870 | 6148494 | ||
X74331.1 | - | 9 | 878 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
