siRNA Details
Gene Symbol: KRTAP10-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198695.2 | NP_941968.2 | 1 | 56 | ||
GenBank mRNA | AB076355.1 | - | 1 | 56 | ||
BC127016.1 | - | 1 | 50 | 40119338 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
