siRNA Details
Gene Symbol: TEKT4P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038327.2 | 4 | 1598 | |||
NR_038328.2 | 4 | 658 | ||||
GenBank mRNA | BC073934.1 | - | 4 | 1589 | 5217232 |
Gene Symbol: MAFIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046439.2 | 8 | 2636 | |||
NR_046440.2 | 5 | 822 | ||||
NR_046441.2 | 3 | 2220 | ||||
GenBank mRNA | DQ786282.1 | - | 5 | 804 |
Gene Symbol: ZNF93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC039001.1 | - | 730 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
