siRNA Details
Gene Symbol: MAFIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046439.2 | 8 | 2513 | |||
NR_046440.2 | 5 | 699 | ||||
NR_046441.2 | 3 | 2097 | ||||
GenBank mRNA | AF289559.1 | - | 3 | 2047 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
