siRNA Details
Gene Symbol: TEKT4P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_038327.2 | 4 | 1596 | |||
NR_038328.2 | 4 | 656 | ||||
GenBank mRNA | BC073934.1 | - | 4 | 1587 | 5217232 |
Gene Symbol: MAFIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_046439.2 | 8 | 2634 | |||
NR_046440.2 | 5 | 820 | ||||
NR_046441.2 | 3 | 2218 | ||||
GenBank mRNA | DQ786282.1 | - | 5 | 802 |
Gene Symbol: ZNF93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC039001.1 | - | 728 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
