siRNA Details
Gene Symbol: LOC100233156 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037871.1 | 5 | 613 | |||
NR_037872.1 | 4 | 1467 | ||||
GenBank mRNA | AK092792.1 | - | 4 | 1448 | ||
BC104658.1 | - | 5 | 580 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
