siRNA Details
Gene Symbol: DPF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001135155.2 | NP_001128627.1 | 3, 4 | 404 | ||
NM_001135156.2 | NP_001128628.1 | 3, 4 | 418 | |||
NM_001289978.1 | NP_001276907.1 | 3, 4 | 404 | |||
NM_004647.3 | NP_004638.2 | 3, 4 | 404 | |||
XM_005259289.2 | XP_005259346.1 | 3, 4 | 404 | |||
XM_005259290.3 | XP_005259347.1 | 3, 4 | 375 | |||
XM_005259292.4 | XP_005259349.1 | 3, 4 | 347 | |||
XM_006723407.2 | XP_006723470.1 | 3, 4 | 375 | |||
XM_006723408.3 | XP_006723471.1 | 3, 4 | 349 | |||
XM_006723409.2 | XP_006723472.1 | 3, 4 | 404 | |||
XM_011527356.1 | XP_011525658.1 | 3, 4 | 404 | |||
XM_011527357.2 | XP_011525659.1 | 3, 4 | 350 | |||
XM_011527358.2 | XP_011525660.1 | 3, 4 | 419 | |||
GenBank mRNA | AK094632.1 | - | 3, 4 | 731 | ||
AK297581.1 | - | 4, 5 | 378 | |||
AK308563.1 | - | 3, 4 | 404 | |||
BC021191.2 | - | 3, 4 | 377 | |||
BC125152.1 | - | 3, 4 | 306 | |||
BC125153.1 | - | 3, 4 | 306 | 40120602 | ||
GQ358728.1 | - | 3, 4 | 296 | |||
U43843.1 | - | 3, 4 | 302 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
