siRNA Details
Gene Symbol: UBE3C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014671.2 | NP_055486.2 | 2 | 408 | ||
XM_005249564.4 | XP_005249621.1 | 2 | 417 | |||
XM_017012818.1 | XP_016868307.1 | 3 | 680 | |||
GenBank mRNA | AK295138.1 | - | 2 | 88 | ||
BC014029.2 | - | 3 | 168 | 4541634 | ||
D13635.1 | - | 2 | 399 | |||
DB454915.1 | - | 2 | 408 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
