siRNA Details
Gene Symbol: PKP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000299.3 | NP_000290.2 | 6 | 1326 | ||
NM_001005337.2 | NP_001005337.1 | 6 | 1326 | |||
GenBank mRNA | AK315291.1 | - | 6 | 1147 | ||
BC114571.1 | - | 6 | 1209 | 40082609 | ||
BC115702.1 | - | 6 | 971 | |||
X79293.1 | - | 6 | 1263 | |||
Z34974.1 | - | 6 | 1327 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
