siRNA Details
Gene Symbol: PKP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000299.3 | NP_000290.2 | 13, 14 | 2426 | ||
NM_001005337.2 | NP_001005337.1 | 12, 13 | 2363 | |||
GenBank mRNA | AK299473.1 | - | 4, 5 | 498 | ||
AK315291.1 | - | 12, 13 | 2184 | |||
AK316131.1 | - | 4, 5 | 498 | |||
BC114571.1 | - | 12, 13 | 2246 | 40082609 | ||
BC115702.1 | - | 12, 13 | 2008 | |||
X79293.1 | - | 12, 13 | 2300 | |||
Z34974.1 | - | 12, 13 | 2364 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
