siRNA Details
Gene Symbol: DNAH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015512.4 | NP_056327.4 | 29 | 5074 | ||
XM_017006129.1 | XP_016861618.1 | 30 | 7962 | |||
XM_017006130.1 | XP_016861619.1 | 30 | 7961 | |||
XM_017006131.1 | XP_016861620.1 | 30 | 7962 | |||
XM_017006132.1 | XP_016861621.1 | 30 | 7963 | |||
XM_017006133.1 | XP_016861622.1 | 30 | 7963 | |||
GenBank mRNA | AB037831.3 | - | 29 | 5074 | ||
AB290163.1 | - | 29 | 5074 | |||
AJ132083.1 | - | 2 | 187 | |||
U61738.1 | - | 72 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
