siRNA Details
Gene Symbol: ACTR3BP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_045000.1 | 1 | 1035 | |||
GenBank mRNA | AY026352.1 | - | 1 | 725 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
