siRNA Details
Gene Symbol: HIST1H4L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003546.2 | NP_003537.1 | 1 | 171 | ||
GenBank mRNA | AB451459.1 | - | 1 | 170 | ||
BC069392.1 | - | 1 | 171 | 7262681 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
