siRNA Details
Gene Symbol: HIST1H2BE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003523.2 | NP_003514.2 | 1 | 403 | ||
GenBank mRNA | BC101748.1 | - | 1 | 470 | 8069254 | |
BC101750.1 | - | 1 | 469 | 8069256 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
