siRNA Details
Gene Symbol: HIST2H2BF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024599.4 | NP_001019770.1 | 1 | 109 | ||
NM_001161334.1 | NP_001154806.1 | 1 | 109 | |||
GenBank mRNA | AK289664.1 | - | 1 | 106 | ||
AK296916.1 | - | 1 | 109 | |||
AK299108.1 | - | 1 | 106 | |||
BC110793.1 | - | 1 | 92 | 5224812 | ||
DA042901.1 | - | 1 | 109 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
