siRNA Details
Gene Symbol: HIST1H4F | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003540.3 | NP_003531.1 | 1 | 144 | ||
GenBank mRNA | BC069288.1 | - | 1 | 144 | 7262669 | |
BC093763.1 | - | 1 | 237 | 7939608 | ||
BC093765.1 | - | 1 | 237 | 7939610 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
