siRNA Details
Gene Symbol: HELZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014877.3 | NP_055692.2 | 4 | 228 | ||
XM_005257888.4 | XP_005257945.1 | 4 | 276 | |||
XM_005257889.4 | XP_005257946.1 | 4 | 276 | |||
XM_006722214.3 | XP_006722277.1 | 4 | 276 | |||
XM_006722215.3 | XP_006722278.1 | 4 | 276 | |||
XM_011525544.2 | XP_011523846.1 | 4 | 347 | |||
XM_017025477.1 | XP_016880966.1 | 4 | 276 | |||
XM_017025478.1 | XP_016880967.1 | 4 | 276 | |||
GenBank mRNA | AK310217.1 | - | 3 | 197 | ||
BC056895.1 | - | 3 | 171 | |||
BC094881.1 | - | 2 | 95 | 3076113 | ||
BC130582.1 | - | 4 | 208 | 40146613 | ||
BC144083.1 | - | 4 | 208 | 9052603 | ||
BP353830.1 | - | 4 | 228 | |||
D29677.2 | - | 3 | 186 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
