siRNA Details
Gene Symbol: HELZ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014877.3 | NP_055692.2 | 13 | 1570 | ||
XM_005257888.4 | XP_005257945.1 | 13 | 1618 | |||
XM_005257889.4 | XP_005257946.1 | 13 | 1618 | |||
XM_006722214.3 | XP_006722277.1 | 13 | 1618 | |||
XM_006722215.3 | XP_006722278.1 | 11 | 1490 | |||
XM_006722216.3 | XP_006722279.1 | 2 | 320 | |||
XM_011525544.2 | XP_011523846.1 | 13 | 1689 | |||
XM_017025477.1 | XP_016880966.1 | 11 | 1490 | |||
XM_017025478.1 | XP_016880967.1 | 14 | 1752 | |||
GenBank mRNA | BC094881.1 | - | 11 | 1437 | 3076113 | |
BC130582.1 | - | 13 | 1550 | 40146613 | ||
BC144083.1 | - | 13 | 1550 | 9052603 | ||
D29677.2 | - | 12 | 1528 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
