siRNA Details
Gene Symbol: LYST | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000081.3 | NP_000072.2 | 3, 4 | 374 | ||
NM_001301365.1 | NP_001288294.1 | 3, 4 | 811 | |||
XM_011544031.1 | XP_011542333.1 | 3, 4 | 374 | |||
XM_011544032.1 | XP_011542334.1 | 3, 4 | 777 | |||
XM_011544033.2 | XP_011542335.1 | 3, 4 | 524 | |||
XM_011544034.1 | XP_011542336.1 | 3, 4 | 374 | |||
XM_011544035.2 | XP_011542337.1 | 3, 4 | 374 | |||
XM_011544037.2 | XP_011542339.1 | 3, 4 | 374 | |||
XM_011544039.2 | XP_011542341.1 | 3, 4 | 374 | |||
XM_011544040.2 | XP_011542342.1 | 3, 4 | 374 | |||
XM_017000150.1 | XP_016855639.1 | 3, 4 | 374 | |||
XM_017000151.1 | XP_016855640.1 | 3, 4 | 374 | |||
GenBank mRNA | AK307457.1 | - | 3, 4 | 777 | ||
BC144702.1 | - | 3, 4 | 675 | |||
L77889.1 | - | 3, 4 | 744 | |||
U67615.1 | - | 3, 4 | 381 | |||
U70064.1 | - | 3, 4 | 744 | |||
U72192.1 | - | 3, 4 | 744 | |||
U84744.1 | - | 3, 4 | 744 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
