siRNA Details
Gene Symbol: ACTR3BP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_027714.1 | 1 | 461 | |||
GenBank mRNA | AY026351.1 | - | 1 | 461 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
