siRNA Details
Gene Symbol: IFRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007245.2 | NP_001007246.1 | 8 | 1158 | ||
NM_001197079.1 | NP_001184008.1 | 7 | 1048 | |||
NM_001197080.1 | NP_001184009.1 | 7 | 883 | |||
NM_001550.3 | NP_001541.2 | 7 | 949 | |||
NR_120333.1 | 6 | 711 | ||||
XM_011516142.2 | XP_011514444.1 | 7 | 944 | |||
XM_011516143.2 | XP_011514445.1 | 7 | 1129 | |||
XM_017012151.1 | XP_016867640.1 | 7 | 1196 | |||
GenBank mRNA | AK297509.1 | - | 6 | 536 | ||
AK298894.1 | - | 7 | 689 | |||
AK308066.1 | - | 8 | 1133 | |||
AK313211.1 | - | 8 | 1158 | |||
AK315967.1 | - | 7 | 881 | |||
BC001272.1 | - | 7 | 902 | 3458309 | ||
BT019889.1 | - | 7 | 688 | |||
BT019890.1 | - | 7 | 688 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
