siRNA Details
Gene Symbol: IFRD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001007245.2 | NP_001007246.1 | 3 | 642 | ||
NM_001197079.1 | NP_001184008.1 | 2 | 532 | |||
NM_001197080.1 | NP_001184009.1 | 2 | 367 | |||
NM_001550.3 | NP_001541.2 | 2 | 433 | |||
NR_120333.1 | 2 | 353 | ||||
XM_011516142.2 | XP_011514444.1 | 2 | 428 | |||
XM_011516143.2 | XP_011514445.1 | 2 | 613 | |||
XM_017012151.1 | XP_016867640.1 | 2 | 680 | |||
GenBank mRNA | AK225222.1 | - | 2 | 406 | ||
AK297509.1 | - | 2 | 178 | |||
AK298894.1 | - | 2 | 173 | |||
AK308066.1 | - | 3 | 617 | |||
AK313211.1 | - | 3 | 642 | |||
AK315967.1 | - | 2 | 365 | |||
AU280361.1 | - | 2 | 430 | |||
BC001272.1 | - | 2 | 386 | 3458309 | ||
BT019889.1 | - | 2 | 172 | |||
BT019890.1 | - | 2 | 172 | |||
CA389949.1 | - | 3 | 615 | |||
CN265021.1 | - | 2 | 353 | |||
DC314572.1 | - | 2 | 530 | |||
DC352698.1 | - | 2 | 433 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
